Learn More
OPA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 27 publications
Brand: Novus Biologicals NB110-55290SS
This item is not returnable.
View return policy
Description
OPA1 Polyclonal specifically detects OPA1 in Human, Mouse, Rat, Porcine, Chicken, Zebrafish samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.
Specifications
| OPA1 | |
| Western Blot 2 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34923139), Immunohistochemistry-Paraffin 2.5 ug/ml, Immunoblotting reported in scientific literature (PMID 25224038) | |
| OPA1 | |
| Pig | |
| IgG |
| Immunocytochemistry | |
| FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 | |
| Affinity Purified | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |