Learn More
OPALIN Antibody (CL11144) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07995AF532
Product Code. 30297523
  • 737.00 € / 0.10mL
Description
OPALIN Monoclonal antibody specifically detects OPALIN in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OPALIN
Monoclonal
Alexa Fluor 532
HTMP10transmembrane protein TMP10, oligodendrocytic myelin paranodal and inner loop proteinTMEM10, TMP10, Transmembrane protein 10opalin
This antibody was developed using a recombinant protein derived from Q96PE5, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL11144
50mM Sodium Borate
Mouse
Protein A purified
RUO
93377
Store at 4⪚C in the dark.
IgG1