missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPRK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 44302G
Additional Details : Weight : 0.50000kg
Description
This gene encodes an opioid receptor, which is a member of the 7 transmembrane-spanning G protein-coupled receptor family. It functions as a receptor for endogenous ligands, as well as a receptor for various synthetic opioids. Ligand binding results in inhibition of adenylate cyclase activity and neurotransmitter release. This opioid receptor plays a role in the perception of pain and mediating the hypolocomotor, analgesic and aversive actions of synthetic opioids. Variations in this gene have also been associated with alcohol dependence and opiate addiction. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. A recent study provided evidence for translational readthrough in this gene, and expression of an additional C-terminally extended isoform via the use of an alternative in-frame translation termination codon.Specifications
OPRK1 | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3 | |
P33534, P34975 | |
OPRK1 | |
The antiserum was produced against a chemically synthesized peptide derived from the internal region of the mouse k-opioid receptor. The sequence is conserved in rat. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
OPRK1 | |
kappa opioid receptor; kappa-type opioid receptor; KOPR1; KOR; K-OR-1; KOR-1; MSL-1; Opiate receptor, kappa-1; opioid receptor kappa 1; opioid receptor, kappa 1; opioid receptor, kappa 2; OPRK; OPRK1; Oprk2; Ork1; R21; Ror-d | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18387, 29335 | |
-20°C | |
Liquid |