Learn More
Thermo Scientific™ Optical Adhesive Film, for Piko PCR plates
Product Code. 15266207 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Sheets
400 Sheets
Unit Size:
100 sheets
Pack of 400
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15266207 100 Sheets 100 sheets
10720181 400 Sheets Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15266207 Supplier Thermo Scientific™ Supplier No. ASF0020100
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
The exceptionally clear Thermo ScientificPiko PCR/qPCR Film Seal allows high transmission of fluorescence across the range of commonly-used fluorophores. When used with aPiko Thermal CyclerorPikoReal Real-Time PCR System, sample volumes as low as 5μL can be used with minimal evaporative loss. These optical adhesive films are available in the ¼-size format for Piko PCR Plates.
When used with a Piko Thermal Cycler or PikoReal Real-Time PCR System, sample volumes as low as 5 µL can be used with minimal evaporative loss. These optical films are available in the ¼-size format for Piko PCR Plates.
Features of the Thermo Scientific Piko PCR/qPCR Film Seals
• Optical clarity with minimal auto-fluorescence for real-time PCR applications
• Tight sealing for sample volumes as low as 5 µL
• Adhesive backing allows easy application
• Cryo-compatible adhesive is effective to –80°C
Related Products
Optical Adhesive Film, for Piko PCR plates
Optical Heat Seal, for Piko PCR plates
TRUSTED_SUSTAINABILITY
Specifications
Color Optical
Quantity 100 Sheets
Product Type Film, Seal, or Applicator
Product Line Piko
Adhesive Pressure Sensitive Optical Seal
Plate Compatibility Piko Plate