missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ d-6 GC Capillary Column

Brand:  Macherey-Nagel™ 726482.60

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10040462

  • 1435.00 € / Each

Estimated Shipment: 24-04-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Optima™ d-6
0.32 mm
Ideal for MSD and PND Detectors
1 µm
Optima™ d-6
GC
GC Capillary Column
0.5 mm
60 m
1 Each
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers