missing translation for 'onlineSavingsMsg'
Learn More

Optimising solution, Elements - Mg,Cu,Rh,Cd,In,Ba,Ce,Pb,U for ICP 0.01mg/L, Fisher Chemical™

Product Code. 12936223 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12936223 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12936223 Supplier Fisher Chemical Supplier No. OP 002 500
  • ONLINE EXCLUSIVE PRICE
    976.22 €
    672.89 € / 500mL
    Save 303.33 €
    31% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Chemical Name or Material Optimising Solution (9 Elements)
Packaging Bottle
Analyte or Component Names Mg,Al,Cr,Mn,Cu,Rh,In,Cd,Ce,Pb,Th,B,Ba
Physical Form Liquid
Concentration 0.01 mg/L
UN Number 1760
Type Optimizing Solution
Element Mg, Al, Cr, Mn, Cu, Rh, In, Cd, Ce, Pb, Th, Ba
Solvent or Matrix Nitric Acid
Quantity 500 mL
compliance-icons
Product Identifier
  • Optimising Solution - 0,01mg/l each of; Mg ; Cu ; Rh ; Cd ; In ; Ba ; ,Ce ; Pb ; U in HNO
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands, forearms andface thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid, copperdinitrate, indium(III) nitrate, pentahydrate, cadmium nitrate, rhodium trichlori
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.