Learn More
SHP Steriltechnik AG Option V Laboklav 25, Vacuum
Product Code. 31908832
Change view
Click to view available options
For Use With (Equipment):
Autoclaves, Laboklav 25, Laboklav ECO
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31908832 Autoclaves, Laboklav 25, Laboklav ECO Each
1 options
Product Code. 31908832 Supplier SHP Steriltechnik AG Supplier No. 21.0000.001.004
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Laboklav 25 series can be optionally equipped with vacuum options.
  • Permanent or fractional pre-vacuum for optimal air removal
  • Safe penetration of steam into sterilisation items that are difficult to vent
  • Reproducible sterilisation results in accordance with DIN 58951-2
  • Drying vacuum for efficient post-drying of packaged items
  • Condensate is reliably removed from the packaging
  • Avoids the need for additional transfer to a drying cabinet
  • Suitable for solid, moist or packaged items
Specifications
For Use With (Equipment) Autoclaves, Laboklav 25, Laboklav ECO
Model Option V Laboklav 25 and ECO
Product Type Autoclave Vacuum