missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR4M1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
288.00 € - 589.00 €
Specifications
| Antigen | OR4M1 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610236
|
Novus Biologicals
NBP2-46853-25ul |
25 μL |
288.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657555
|
Novus Biologicals
NBP2-46853 |
0.1 mL |
589.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OR4M1 Polyclonal antibody specifically detects OR4M1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| OR4M1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8NGD0 | |
| 441670 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| olfactory receptor 4M1, Olfactory receptor OR14-7, olfactory receptor, family 4, subfamily M, member 1, OR14-7 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: NELDSYFCDITQVVRIACANTFPEELV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title