missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR5D13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06140-50ul
This item is not returnable.
View return policy
Description
OR5D13 Polyclonal antibody specifically detects OR5D13 in Human samples. It is validated for Western Blot
Specifications
| OR5D13 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| olfactory receptor 5D13, Olfactory receptor OR11-142, Olfactory receptor OR11-148, olfactory receptor, family 5, subfamily D, member 13 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human OR5D13. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 390142 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur