missing translation for 'onlineSavingsMsg'
Learn More

Orantinib, MedChemExpress

Product Code. 16474018
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474018 10 mg 10mg
16420377 50 mg 50mg
16415016 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474018 Supplier Medchem Express Supplier No. HY10517
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Orantinib (SU6668; TSU-68) is a multi-targeted receptor tyrosine kinase inhibitor with Kis of 2.1 μM, 8 nM and 1.2 μM for Flt-1, PDGFRβ and FGFR1, respectively.

Orantinib (SU6668

Specifications

Chemical Name or Material Orantinib
Physical Form Powder
CAS 252916-29-3
Color Deep Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H18N2O3
For Use With (Application) Cancer-Kinase/protease
Synonym SU6668 TSU-68
Solubility Information DMSO : ≥ 28 mg/mL (90.22 mM)
SMILES CC1=C(/C=C2C3=CC=CC=C3NC/2=O)NC(C)=C1CCC(O)=O
Molecular Weight (g/mol) 310.35
Formula Weight 310.35
Percent Purity 98.18%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.