missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24600SS
This item is not returnable.
View return policy
Description
ORC1 Polyclonal specifically detects ORC1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| ORC1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ORC1 | |
| A portion of amino acids 170 to 220 of human ORC1L was used as immunogen. | |
| 0.025 mg | |
| Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4998 | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml | |
| HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, 89% homologous in dog and 84% homologous in mouse and rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction