Learn More
ORC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17784-1-AP-20UL
 View more versions of this product
Product Code. 16837764
  • 145.00 € / 20µL
Description
The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. Gene silencing studies with small interfering RNA demonstrated that this protein plays an essential role in coordinating chromosome replication and segregation with cytokinesis.
Specifications
Specifications
ORC6
Polyclonal
Unconjugated
ORC6
ORC6, ORC6L
Rabbit
Antigen Affinity Chromatography
RUO
23594, 291927, 56452
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9WUJ8, Q9Y5N6
Orc6
ORC6 Fusion Protein Ag11769
20 μL
Primary
Rat, Mouse, Human