missing translation for 'onlineSavingsMsg'
Learn More

Orcinol, 98%

Product Code. p-4229318 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10637211

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-15-4 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-15-4
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002291
InChI Key OIPPWFOQEKKFEE-UHFFFAOYSA-N
Synonym orcinol, 3,5-dihydroxytoluene, 5-methylresorcinol, 1,3-dihydroxy-5-methylbenzene, orcin, 5-methyl-1,3-benzenediol, 5-methylresorcin, 3-hydroxy-5-methylphenol, 3,5-toluenediol, 1,3-benzenediol, 5-methyl
PubChem CID 10436
ChEBI CHEBI:16536
IUPAC Name 5-methylbenzene-1,3-diol
SMILES CC1=CC(O)=CC(O)=C1

Specifications

Melting Point 105.0°C to 110.0°C
Color Brown-Pink to Gray-Pink
Boiling Point 290.0°C
Flash Point 159°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,6959
Formula Weight 124.14
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Orcinol
Show More Show Less
compliance-icons
Product Identifier
  • Orcinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/in accordance with local/regional/national/international regulations..
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.