missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oregon Pesticide Mixture 2-100 AL 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | (E)-Fenpyroximate∣Dimethoate∣Ethoprophos∣Etofenprox∣Etoxazole∣Fenoxycarb∣Fipronil∣Flonicamid∣Fludioxonil∣Hexythiazox∣Imazalil∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methiocarb∣Methomyl∣MGK 264 isomer A∣MGK 264 isomer B∣Myclobutanil∣Parathion-methyl |
| CAS | 134098-61-6, 60-51-5, 13194-48-4, 80844-07-1, 153233-91-1, 72490-01-8 |
| Analyte or Component Names | (E)-Fenpyroximate∣Dimethoate∣Ethoprophos∣Etofenprox∣Etoxazole∣Fenoxycarb∣Fipronil∣Flonicamid∣Fludioxonil∣Hexythiazox∣Imazalil∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methiocarb∣Methomyl∣MGK 264 isomer A∣MGK 264 isomer B∣Myclobutanil∣Parathion-methyl |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | MGK 264 isomer B 30 μg/mL∣MGK 264 isomer A 70 μg/mL∣(E)-Fenpyroximate 100 μg/mL∣Dimethoate 100 μg/mL∣Ethoprophos 100 μg/mL∣Etofenprox 100 μg/mL∣Etoxazole 100 μg/mL∣Fenoxycarb 100 μg/mL∣Fipronil 100 μg/mL∣Flonicamid 100 μg/mL∣Fludioxonil 100 μg/mL∣Hexythiazox 100 μg/mL∣Imazalil 100 μg/mL∣Imidacloprid 100 μg/mL∣Kresoxim-methyl 100 μg/mL∣Malathion 100 μg/mL∣Metalaxyl 100 μg/mL∣Methiocarb 100 μg/mL∣Methomyl 100 μg/mL∣Myclobutanil 100 μg/mL∣Parathion-methyl 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?