missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oregon Pesticide Mixture 4 600 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Carbaryl∣Carbofuran∣Chlorantraniliprole∣Clofentezine∣Daminozide∣Fenoxycarb∣Imazalil∣Myclobutanil∣Paclobutrazol∣Propiconazole∣Propoxur∣Tebuconazole |
| CAS | 63-25-2, 1563-66-2, 500008-45-7, 74115-24-5, 1596-84-5, 72490-01-8 |
| Analyte or Component Names | Carbaryl∣Carbofuran∣Chlorantraniliprole∣Clofentezine∣Daminozide∣Fenoxycarb∣Imazalil∣Myclobutanil∣Paclobutrazol∣Propiconazole∣Propoxur∣Tebuconazole |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Carbaryl 600 μg/mL∣Carbofuran 600 μg/mL∣Chlorantraniliprole 600 μg/mL∣Clofentezine 600 μg/mL∣Daminozide 600 μg/mL∣Fenoxycarb 600 μg/mL∣Imazalil 600 μg/mL∣Myclobutanil 600 μg/mL∣Paclobutrazol 600 μg/mL∣Propiconazole 600 μg/mL∣Propoxur 600 μg/mL∣Tebuconazole 600 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?