missing translation for 'onlineSavingsMsg'
Learn More

Oregon Pesticide Mixture 662 100x Action Limit 100-200 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30311700
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311700 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311700 Supplier Dr. Ehrenstorfer Supplier No. DREA50000662AL
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acequinocyl∣Chlorfenapyr∣Cyfluthrin∣Cypermethrin∣Daminozide∣Dichlorvos∣Flonicamid∣Hexythiazox∣Oxamyl∣Piperonyl butoxide∣Pyrethrins
CAS 57960-19-7, 122453-73-0, 68359-37-5, 52315-07-8, 1596-84-5, 62-73-7
Analyte or Component Names Acequinocyl∣Chlorfenapyr∣Cyfluthrin∣Cypermethrin∣Daminozide∣Dichlorvos∣Flonicamid∣Hexythiazox∣Oxamyl∣Piperonyl butoxide∣Pyrethrins
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Chlorfenapyr 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Daminozide 100 μg/mL∣Dichlorvos 100 μg/mL∣Flonicamid 100 μg/mL∣Hexythiazox 100 μg/mL∣Oxamyl 100 μg/mL∣Pyrethrins 100 μg/mL∣Acequinocyl 200 μg/mL∣Piperonyl butoxide 200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.