missing translation for 'onlineSavingsMsg'
Learn More

Orexin 2 Receptor Agonist, MedChemExpress

Product Code. p-200101283
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477586 100 mg 100mg
16463697 50 mg 50mg
16440188 5 mg 5mg
16417728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477586 Supplier Medchem Express Supplier No. HY19320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Orexin 2 Receptor Agonist is a potent (EC50 on OX2R is 23 nM) and OX2R-selective (OX1R/OX2R EC50 ratio is 70) agonist.

Orexin 2 Receptor Agonist is a potent (EC50 on OX2R is 23 nM) and OX2R-selective (OX1R/OX2R EC50 ratio is 70) agonist.

Specifications

Chemical Name or Material Orexin 2 Receptor Agonist
CAS 1796565-52-0
Color White
Purity Grade Notes Research
Molecular Formula C32H34N4O5S
Quantity 100 mg
Solubility Information DMSO : ≥ 32 mg/mL (54.54 mM)
SMILES O=C(C1=CC=CC(C)=C1)NCCNC2=CC(NS(C3=C(OC)C=CC(C4=CC(C(N(C)C)=O)=CC=C4)=C3)(=O)=O)=CC=C2
Molecular Weight (g/mol) 586.7
Formula Weight 586.7
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.