missing translation for 'onlineSavingsMsg'
Learn More

Org 27569, Tocris Bioscience™

Product Code. 18055982
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055982 10 mg 10mg
18004202 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18055982 Supplier Tocris Bioscience Supplier No. 2957/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent allosteric modulator of CB1 receptors

Potent CB1 receptor allosteric modulator (pEC50 = 8.24). Significantly increases binding of the CB1 agonist [3H]CP 55.940 (pKb = 5.67) and decreases binding of the CB1 inverse agonist [3H]SR 141716A (pKb = 5.95). Inhibits CB1 receptor antagonist efficacy in vitro (pKb = 7.57).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868273-06-7
Molecular Formula C24H28ClN3O
Molecular Weight (g/mol) 409.96
MDL Number MFCD11045300
InChI Key AHFZDNYNXFMRFQ-UHFFFAOYSA-N
Synonym 5-chloro-3-ethyl-n-4-piperidin-1-yl phenethyl-1h-indole-2-carboxamide, 5-chloro-3-ethyl-1h-indole-2-carboxylic acid 2-4-piperidin-1-yl phenylethyl amide, 5-chloro-3-ethyl-n-2-4-piperidin-1-yl phenyl ethyl-1h-indole-2-carboxamide, d05qtx, org hplc, 5-chloro-3-ethyl-1h-indole-2-carboxylic acid 2-4-piperidin-1-ylphenyl ethyl amide, 5-chloro-3-ethyl-n-2-4-piperidin-1-ylphenyl ethyl-1h-indole-2-carboxamide, 5-chloro-3-ethyl-n-2-4-1-piperidinyl phenyl ethyl-1h-indole-2-carboxamide
PubChem CID 44828492
IUPAC Name 5-chloro-3-ethyl-N-{2-[4-(piperidin-1-yl)phenyl]ethyl}-1H-indole-2-carboxamide
SMILES CCC1=C(NC2=CC=C(Cl)C=C12)C(=O)NCCC1=CC=C(C=C1)N1CCCCC1

Specifications

Quantity 10 mg
Formula Weight 409.95
Percent Purity >99%
Chemical Name or Material Org 27569
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.