missing translation for 'onlineSavingsMsg'
Learn More

Organochlorine Pesticides Mixture 921 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30319118
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319118 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319118 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000921AC
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor
CAS 72-54-8, 72-55-9, 50-29-3, 309-00-2, 959-98-8, 319-84-6
Analyte or Component Names 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 4,4'-DDD 2000 μg/mL∣4,4'-DDE 2000 μg/mL∣4,4'-DDT 2000 μg/mL∣Aldrin 2000 μg/mL∣alpha-Endosulfan 2000 μg/mL∣alpha-HCH 2000 μg/mL∣beta-Endosulfan 2000 μg/mL∣beta-HCH 2000 μg/mL∣delta-HCH 2000 μg/mL∣Dieldrin 2000 μg/mL∣Endosulfan-sulfate 2000 μg/mL∣Endrin 2000 μg/mL∣Endrin-aldehyde 2000 μg/mL∣Endrin-ketone 2000 μg/mL∣gamma-HCH 2000 μg/mL∣Heptachlor 2000 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 2000 μg/mL∣Methoxychlor 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.