missing translation for 'onlineSavingsMsg'
Learn More

Organometallic Oil Standard, 21-Element, 500μg/g, SPEX CertiPrep™

Product Code. 11460349 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460349 50 g 50g
11450349 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460349 Supplier SPEX Certiprep Supplier No. S21500Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag500μg/g
Al500μg/g
B500μg/g
Ba500μg/g
Ca500μg/g
Cd500μg/g
Cr500μg/g
Cu500μg/g
Fe500μg/g
Mg500μg/g
Mn500μg/g
Mo500μg/g
Na500μg/g
Ni500μg/g
P500μg/g
Pb500μg/g
Si500μg/g
Sn500μg/g
Ti500μg/g
V500μg/g
Zn500μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material 21 Element Oil Standard
Packaging Triple-leached LDPE bottle
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Base Oil 75
Industry Type Petrochemical Testing
Quantity 50 g
Grade Oil
Concentration or Composition (by Analyte or Components) Each component at 100μg/g (S21-100), 300μg/g (S21-300), 500μg/g (S21-500), or 900μg/g (S21-900) in Base Oil 75:
Aluminum
Barium
Boron
Cadmium
Calcium
Chromium
Copper
Iron
Lead
Magnesium
Manganese
Molybdenum
Nickel
Phosphorous
Silicon
Silver
Sodium
Tin
Titanium
Vanadium
Zinc
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.