missing translation for 'onlineSavingsMsg'
Learn More

Organophosphorus Pesticides Mixture 624 1000 μg/mL in Methyl tert Butyl Ether, Dr. Ehrenstorfer

Product Code. 30328311
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328311 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328311 Supplier Dr. Ehrenstorfer Supplier No. DREA50000624MB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Demeton (O+S)∣Dichlorvos∣Dimethoate∣Malathion∣Methamidophos∣Parathion-ethyl∣Parathion-methyl∣Trichlorphon
CAS 8065-48-3, 62-73-7, 60-51-5, 121-75-5, 10265-92-6, 56-38-2
Analyte or Component Names Demeton (O+S)∣Dichlorvos∣Dimethoate∣Malathion∣Methamidophos∣Parathion-ethyl∣Parathion-methyl∣Trichlorphon
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix MTBE
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Demeton (O+S) 1000 μg/mL∣Dichlorvos 1000 μg/mL∣Dimethoate 1000 μg/mL∣Malathion 1000 μg/mL∣Methamidophos 1000 μg/mL∣Parathion-ethyl 1000 μg/mL∣Parathion-methyl 1000 μg/mL∣Trichlorphon 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.