Learn More
Thermo Scientific™ Orion™ 2100 Series and 1800 Series Analyzer Reagents for Orion™ Online Monitors
Product Code. 15837134
Change view
Click to view available options
Type:
Formic Acid Reagent
Unit Size:
Each
Product Code. Type Unit Size
15837134 Formic Acid Reagent Each
1 options
Product Code. 15837134 Supplier Thermo Scientific™ Supplier No. 182011
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
Denne vare kan ikke returneres. Se returpolicy
Use these analyzer reagents for our process analyzers.
Use Thermo Scientific™ Orion™ 2100 Series and 1800 Series Analyzer Reagents for Orion™ Online Monitors with Orion™ process analyzers.
Reagents for the Orion 2109XP, 2110XP, 2111LL, 2111XP, 2117XP, 2118XP, 2120XP, 1809, 1810, 1811AO, 1811EL, 1817HL, 1817LL, 1818 and 1820 analyzers.
TRUSTED_SUSTAINABILITY
Tekniske data
For Use With (Equipment) 2109XP Fluoride, 2117XP Chloride and 2120XP Calcium Hardness Analyzers
Type Formic Acid Reagent
Quantity 1 L
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.