missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ORP Standards for Redox/ORP Electrodes

Product Code. 11727697
Change view
Click to view available options
Quantity:
475 mL
5 x 60 mL
Unit Size:
475mL
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11727697 475 mL 475mL
11757807 5 x 60 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11727697 Supplier Thermo Scientific™ Supplier No. 967901
  • ONLINE EXCLUSIVE PRICE
    75.25 €
    72.70 € / 475mL
    Save 2.55 €
    3% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measure ORP simply and accurately with these standards, stable for up to a year (unopened).

Measure ORP simply and accurately with Thermo Scientific™ Orion™ ORP Standards for Redox/ORP Electrodes. Orion OPR standards are stable for up to a year (unopened) and can be used directly from the bottle with no mixing or diluting.

Orion OPR standards are designed for use with gel-filled ORP/ATC Triode™ electrodes (9179BNMD and 9179BN); refillable ORP/ATC Triode electrodes (9180BNMD and 9180BN); glass-bodied ORP electrodes, (9778BNWP and 9778SC); and Sure-Flow™ ORP electrodes, (9678BNWP and 967800).

  • Included in the new ASTM ORP method
  • Used as relative mV standard to calibrate the offset of sample
  • +220mV vs. Ag/AgCl using 4M KCl saturated with AgCl filling solution (Part No. 900011), +420mV vs. Standard Hydrogen Electrode (EH)
TRUSTED_SUSTAINABILITY

Specifications

Description ORP Standard
Packaging Bottle
For Use With (Equipment) Gel-filled ORP/ATC Triode™ electrodes (9179BNMD and 9179BN), Refillable ORP/ATC Triode electrodes (9180BNMD and 9180BN), Glass-bodied ORP electrodes, (9778BNWP and 9778SC), Sure-Flow™ ORP electrodes, (9678BNWP and 967800)
Solution Type Standard Solution
Type Redox/ORP Electrode Standard
Quantity 475 mL
compliance-icons
Product Identifier
  • ORP Standard
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P321-Specific treatment (see supplemental first aid instructions on this label).
  • P202-Do not handle until all safety precautions have been read and understood.
Supplemental information
  • MIXTURE LIST-Contain: Potassium Iodide, Potassium Hydroxide, Iodine, Boric Acid

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.