missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Buffer Bottles

Product Code. 11797797
Change view
Click to view available options
Quantity:
475 mL
5 x 60 mL
pH:
1.68 ± 0.01 pH
10.01 ± 0.02 pH
12.46 ± 0.03 pH
4.01 ± 0.01 pH
5.00 ± 0.02 pH
6.86 ± 0.01 pH
7.00 ± 0.01 pH
9.18 ± 0.01 pH
Unit Size:
475mL
Pack of 5
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH unitSize
11797797 5 x 60 mL 7.00 ± 0.01 pH Pack of 5
10520164 475 mL 1.68 ± 0.01 pH 475mL
11727807 5 x 60 mL 1.68 ± 0.01 pH Pack of 5
10213021 475 mL 10.01 ± 0.02 pH 475mL
10136470 5 x 60 mL 10.01 ± 0.02 pH Pack of 5
10114234 475 mL 12.46 ± 0.03 pH 475mL
11717807 5 x 60 mL 12.46 ± 0.03 pH Pack of 5
10759161 475 mL 4.01 ± 0.01 pH 475mL
11787797 5 x 60 mL 4.01 ± 0.01 pH Pack of 5
11772797 475 mL 5.00 ± 0.02 pH 475mL
11762817 475 mL 6.86 ± 0.01 pH 475mL
11737807 5 x 60 mL 6.86 ± 0.01 pH Pack of 5
10653951 475 mL 7.00 ± 0.01 pH 475mL
11722827 475 mL 9.18 ± 0.01 pH 475mL
11747807 5 x 60 mL 9.18 ± 0.01 pH Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 11797797 Supplier Thermo Scientific™ Supplier No. 910760

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Perform accurate, reproducible pH calibrations with these color-coded buffer bottles.

Use Thermo Scientific™ Orion™ pH buffer bottles to perform accurate, reproducible pH calibrations on your pH meter and pH electrode measurement system. Orion pH buffers are available in 475mL bottles for everyday use and laboratory and bench pH analysis and available in 60 mL bottle 5 packs for occasional use and portable and field pH analysis. Using high-quality pH buffers to calibrate your pH system is the best way to ensure your pH measurements are correct and repeatable.

All Orion pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability. Manufactured under ISO 9000 quality standards.

Buffer Color Codes:

  • pH 4.01 buffer is pink
  • pH 5.00 buffer is orange
  • pH 7.00 buffer is yellow
  • pH 10.01 buffer is blue

The buffers are available in pint (475mL) bottles, portable five packs of 2 oz. (5 x 60mL) bottles, convenient All-in-One buffer kits, 5 gallon (19L) Cubitainers™ and single-use pouches.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material pH Buffer, bottles
Color Yellow
Description Orion pH 7.00 buffer, 5 x 60 mL
For Use With (Application) Accurate pH calibrations and NIST traceability, laboratory and bench analysis
For Use With (Equipment) pH Meter and Electrode
pH 7.00 ± 0.01 pH
Physical Form Liquid
Quantity 5 x 60 mL
Solution Type pH Buffer
Formulation Water, Dibasic, Potassium Dihydrogen Phosphate, 5-Bromo-5-Nitro-1,3-Dioxane, FD & C Yellow #5, Sodium Phosphate
Packaging Bottle
Traceability to NIST Yes
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.