missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches

Código de producto. 10283061
Cambiar vista
Click to view available options
Quantity:
10/Pk.
25/Pk.
pH:
4.01
7
10.01
Tamaño de la unidad:
Pack of 10
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity pH Tamaño de la unidad
10283061 10/Pk. 4.01 Pack of 10
10756861 25/Pk. 4.01 Pack of 25
10069110 10/Pk. 7 Pack of 10
10242491 25/Pk. 7 Pack of 25
10185062 10/Pk. 10.01 Pack of 10
10470443 25/Pk. 10.01 Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10283061 Proveedor Thermo Scientific™ N.º de proveedor 910410WA
 Restricted Item
In Stock
Añadir a la cesta
Edge
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Orion™ single-use pH calibration buffers deliver NIST-traceable accuracy for pH meters and electrodes. Designed for GxP labs, these ready-to-use, color-coded buffer pouches support compliant workflows, reduce contamination risk and ensure reliable, traceable pH calibration results.

Thermo Scientific™ Orion™ single-use pH calibration buffers deliver NIST-traceable accuracy for pH meters and electrodes in laboratory and GxP-regulated environments. These ready-to-use, color-coded buffer pouches support compliant calibration workflows, reduce contamination risk, and ensure consistent, reliable results. Each pouch is individually labeled for full traceability and designed for direct electrode use, improving efficiency and confidence in pH measurements.

Single-Use NIST-Traceable pH Calibration Buffers for pH Meter and Electrode Calibration

Each pouch is individually labeled with catalog number, lot code and expiration date, supporting documentation and traceability required for GMP, GLP and ISO quality systems.

pH 4, 7, and 10 buffer pouches and solutions are color-coded for quick identification during calibration.

  • pH 4 buffer pouches and solution are light red
  • pH 7 buffer pouches and solution are yellow
  • pH 10 buffer pouches and solution are blue

Redesigned pouches offer a rectangular shape to better accommodate electrodes placed directly into the pouch and 20 mL solution volume to ensure proper electrode immersion for accurate calibration.

Key Features

  • pH buffers are traceable to NIST reference standards
  • Each pH buffer pouch and solution is color-coded
  • Each pouch is labeled with catalog number, lot code and expiration date
  • Each pouch is easy to open with tear notches
  • Each pouch is single use to reduce contamination risk

Workflow and Application Benefits

Orion pH buffer individual-use pouches simplify pH calibration procedures in both laboratory and field settings.

  • Direct electrode calibration: Updated pouch design allows electrodes to be placed directly into the pouch for quick calibration steps.
  • Quick identification during calibration: Color-coded buffers help technicians rapidly identify the correct standard during multi-point calibration procedures.
  • Consistent calibration performance: Single-use pouches ensure each calibration uses fresh buffer solution, reducing variability caused by contamination or evaporation.
  • Convenient calibration anywhere: Compact pouches are ideal for field testing, environmental monitoring and routine QA/QC calibration workflows.
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material pH Buffer, individual use pouches
Boiling Point 100°C
Color Pink
Certificate of Analysis Yes
For Use With (Application) pH calibration
For Use With (Equipment) pH Meter and Electrode, single-use
pH 4.01
Physical Form Liquid
Quantity 10/Pk.
Solution Type Buffer
Formulation Water, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate
Packaging Pouch
Traceability to NIST Yes
Solubility Information Soluble
Certifications/Compliance ISO 9001
Grade Certified
Mostrar más Mostrar menos

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.