missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Carbamoyltransferase Antibody (CL4049) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59040APC
This item is not returnable.
View return policy
Description
Ornithine Carbamoyltransferase Monoclonal antibody specifically detects Ornithine Carbamoyltransferase in Human samples. It is validated for Western Blot
Specifications
| Ornithine Carbamoyltransferase | |
| Monoclonal | |
| APC | |
| EC 2.1.3, EC 2.1.3.3, MGC129967, MGC129968, MGC138856, OCTD, ornithine carbamoyltransferase, ornithine carbamoyltransferase, mitochondrial, Ornithine transcarbamylase, OTCase | |
| This antibody was developed using a recombinant protein derived from P00480, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| metabolism | |
| 5009 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL4049 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction