missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP buffer +475mV, Hamilton™
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Specifications
Specifications
| Color | Yellow |
| pH | 1.1 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Buffer |
| Formulation | 2% Diammonium iron bis(sulphate)-6-hydrate, 0.5% Hydrochloric acid |
| Odor | Odorless |
| Solubility Information | Water Soluble |
Safety and Handling
Product Identifier
- Redox Buffer 478
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction