missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562725
This item is not returnable.
View return policy
Description
Immunogen sequence: DQLRDMHQGS ELSRMGVSEA STGQRRLHSL STSSDTTADS FSSLNPEEQE ALYMKGRELT PQLSQTSILS LADSHTEF Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding the same isoform have been identified.
Specifications
| ORP7 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL7 | |
| ORP7; ORP-7; OSBPL7; OSBP-related protein 7; oxysterol binding protein like 7; oxysterol binding protein-like 7; oxysterol-binding protein-related protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114881 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZF2 | |
| OSBPL7 | |
| Recombinant protein corresponding to Human ORP7. Recombinant protein control fragment (Product #RP-103145). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction