missing translation for 'onlineSavingsMsg'
Learn More

Osalmid, MedChemExpress

Product Code. 16492369
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 16492369

Brand: Medchem Express HYB2116

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Osalmid is a ribonucleotide reductase small subunit M2 (RRM2) targeting compound; suppresses ribonucleotide reductase activity with an IC50 of 8.23 μM.

Osalmid is a ribonucleotide reductase small subunit M2 (RRM2) targeting compound

Specifications

Chemical Name or Material Osalmid
CAS 526-18-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H11NO3
Quantity 1 g
Synonym Oxaphenamide 4'-Hydroxysalicylanilide
Solubility Information DMSO : ≥ 100 mg/mL (436.24 mM)
SMILES O=C(NC1=CC=C(O)C=C1)C2=CC=CC=C2O
Molecular Weight (g/mol) 229.23
Formula Weight 229.23
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.