missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteopontin/OPN Antibody (1B20) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 22 publications
244.00 € - 533.00 €
Specifications
| Antigen | Osteopontin/OPN |
|---|---|
| Clone | 1B20 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Simple Western 1:20, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18757853
|
Novus Biologicals
NB110-89062SS |
0.025 mL |
244.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18705693
|
Novus Biologicals
NB110-89062 |
0.1 mL |
533.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Osteopontin/OPN | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia | |
| p10451, P08721 | |
| 6696 | |
| Synthetic peptide corresponding to the C-terminus of human Osteopontin [Swiss-Prot# P10451]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1B20 | |
| Western Blot 1:1000, Simple Western 1:20, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Rabbit | |
| BNSP, Bone sialoprotein 1, MGC110940, Nephropontin, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
| SPP1 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title