missing translation for 'onlineSavingsMsg'
Learn More

Osteopontin/OPN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-89952-25ul

 View more versions of this product

Product Code. 18463792

  • 234.00 € / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Osteopontin/OPN Polyclonal antibody specifically detects Osteopontin/OPN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Osteopontin/OPN
Polyclonal
Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500
BNSP, Bone sialoprotein 1, MGC110940, Nephropontin, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin
This antibody was developed against Recombinant Protein corresponding to amino acids: SQDSIDSNDSDDVDDTDDSHQSDESHHSDESDELVTDFPTDLPATEVFTPVVPTVDTYDGRGDSVVYGLRSKSKKFRRPDIQYPDAT
25 μL
Apoptosis, Cancer, Cellular Markers, Extracellular Matrix, Hypoxia
6696
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.