Learn More
Osteopontin Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30200-1-AP-150UL
Description
The Osteopontin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse Osteopontin. This antibody recognizes human, mouse, rat antigen. The Osteopontin antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| Osteopontin | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| AA960535, AI790405, Apl 1, BNSP, Bsp, BSPI, Eta, ETA 1, minopontin, OP, Opn, Opnl, Osteopontin, Ric, secreted phosphoprotein 1, Spp 1, Spp1 | |
| 150 μL | |
| 20750 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |