missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OTC Recombinant Rabbit Monoclonal Antibody (6B6N2)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Specifications
| Antigen | OTC |
|---|---|
| Clone | 6B6N2 |
| Concentration | 1.2 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30282742
|
Invitrogen™
MA558165 |
20 μL |
176.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30282745
![]() |
Invitrogen™
MA558166 |
100 μL |
436.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: LQAATPKGYE PDASVTKLAE QYAKENGTKL LLTNDPLEAA HGGNVLITDT WISMGQEEEK KKRLQAFQGY QVTMKTAKVA ASDWTFLHCL PRKPEEVDDE VFYSPRSLVF PEAENRKWTI MAVMVSLLTD YSPQLQKPKF.
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.Specifications
| OTC | |
| 1.2 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P00480 | |
| 5009 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 215-354 of human OTC (NP_000522.3). | |
| Antibody |
| 6B6N2 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| OTC | |
| AI265390; OCTD; ornithine carbamoyltransferase; ornithine carbamoyltransferase, mitochondrial; Ornithine transcarbamylase; Otc; OTCase; Sf; sparse fur; spf | |
| OTC | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title