missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), Brilliant Violet™ 650, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ 416-5743-80

 View more versions of this product

Product Code. 17922001

  • 188.00 € / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal antibody specifically detects OVA257-264 (SIINFEKL) peptide bound to H-2Kb in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OVA257-264 (SIINFEKL) peptide bound to H-2Kb
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
Mouse
25 μg
Primary
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
eBio25-D1.16 (25-D1.16)
Brilliant Violet 650
H-2Kb-SIINFEKL; OVA257-264; OVA257-264 H-2Kb; OVA-Kb
Affinity chromatography
RUO
Mouse
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.