missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Ovary Tissue Slides (Normal)- Paraffin

Product Code. 18061464 Shop All Bio Techne Products
Change view
Click to view available options
Ovary Tissue Slides (Normal)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18061464 Ovary Tissue Slides (Normal)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18061464 Supplier Novus Biologicals™ Supplier No. NBP230190
  • ONLINE EXCLUSIVE PRICE
    238.00 €
    224.70 € / Pack of 5
    Save 13.30 €
    5% Off
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Type Ovary Tissue Slides (Normal)
For Use With (Application) Immunohistochemistry-Paraffin
Regulatory Status This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species Human
Tissue Condition Normal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.