missing translation for 'onlineSavingsMsg'
Learn More

Oxalate Standard For Ion Chromatography, SPEX CertiPrep™

Product Code. 15658225 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15658225 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15658225 Supplier SPEX Certiprep Supplier No. ASC2O492Y
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
C2O4-21000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 62-76-0, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 99.9
Analyte or Component Names Disodium oxalate,distilled water,conductivity or of similar purity
Molecular Formula C2Na2O4
MDL Number MFCD00012465
Technique Ion Chromatography
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Type Certified Reference Material
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O
Element Disodium Oxalate
IUPAC Name disodium;oxalate
Solvent or Matrix Water
Molecular Weight (g/mol) 134.00
PubChem CID 6125
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material Oxalate Standard, For Ion Chromatography
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.