missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxaliplatin, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Description
Oxaliplatin is a DNA synthesis inhibitor. Oxaliplatin causes DNA crosslinking damage, prevents DNA replication and transcription and induces apoptosis. Oxaliplatin can be used for cancer research.
Specifications
Specifications
| Chemical Name or Material | Oxaliplatin |
| Physical Form | Solid |
| CAS | 61825-94-3 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C8H14N2O4Pt |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 20.83 mg/mL (52.43 mM; ultrasonic and warming and heat to 60°C; DMSO can inactivate Oxaliplatin's activity) ∣H2O : 2.17 mg/mL (5.46 mM; ultrasonic and warming and heat to 60°C; DMSO can inactivate Oxaliplatin's activity) ∣DMF : 1.67 mg/mL (4.20 mM; Need ultrasonic; DMSO can inactivate Oxaliplatin's activity) ∣Ethanol : < 1 mg/mL (insoluble; DMSO can inactivate Oxaliplatin's activity) |
| SMILES | O=C(O1)C(O[Pt]21[NH2][C@@H]3CCCC[C@H]3[NH2]2)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction