missing translation for 'onlineSavingsMsg'
Learn More

Oxaliplatin, MedChemExpress

Product Code. 16451836
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451836 500 mg 500mg
16443327 50 mg 50mg
16441246 200 mg 200mg
16417266 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16451836 Supplier Medchem Express Supplier No. HY17371

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxaliplatin is a DNA synthesis inhibitor. Oxaliplatin causes DNA crosslinking damage, prevents DNA replication and transcription and induces apoptosis. Oxaliplatin can be used for cancer research.

Oxaliplatin is a DNA synthesis inhibitor. Oxaliplatin causes DNA crosslinking damage, prevents DNA replication and transcription and induces apoptosis. Oxaliplatin can be used for cancer research.

Specifications

Chemical Name or Material Oxaliplatin
Physical Form Solid
CAS 61825-94-3
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C8H14N2O4Pt
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 20.83 mg/mL (52.43 mM; ultrasonic and warming and heat to 60°C; DMSO can inactivate Oxaliplatin's activity) ∣H2O : 2.17 mg/mL (5.46 mM; ultrasonic and warming and heat to 60°C; DMSO can inactivate Oxaliplatin's activity) ∣DMF : 1.67 mg/mL (4.20 mM; Need ultrasonic; DMSO can inactivate Oxaliplatin's activity) ∣Ethanol : < 1 mg/mL (insoluble; DMSO can inactivate Oxaliplatin's activity)
SMILES O=C(O1)C(O[Pt]21[NH2][C@@H]3CCCC[C@H]3[NH2]2)=O
Molecular Weight (g/mol) 397.29
Formula Weight 397.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.