missing translation for 'onlineSavingsMsg'
Learn More

Oxamflatin, TRC

Product Code. 30415175
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30415175 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415175 Supplier Toronto Research Chemicals Supplier No. TRCO8450905MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151720-43-3
Molecular Formula C17 H14 N2 O4 S
Molecular Weight (g/mol) 342.37
IUPAC Name (E)-5-[3-(benzenesulfonamido)phenyl]-N-hydroxypent-2-en-4-ynamide
SMILES ONC(=O)\C=C\C#Cc1cccc(NS(=O)(=O)c2ccccc2)c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H14N2O4S/c20-17(18-21)12-5-4-7-14-8-6-9-15(13-14)19-24(22,23)16-10-2-1-3-11-16/h1-3,5-6,8-13,19,21H,(H,18,20)/b12-5+
Formula Weight 342.0674
Chemical Name or Material Oxamflatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.