missing translation for 'onlineSavingsMsg'
Learn More

Oxaprozin, TRC

Product Code. 30138561
Change view
Click to view available options
Quantity:
5 mg
50 mg
1 g
Unit Size:
1g
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138561 1 g 1g
30137487 50 mg 50mg
17914075 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138561 Supplier Toronto Research Chemicals Supplier No. TRCO8454001G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21256-18-8
Molecular Formula C18 H15 N O3
Molecular Weight (g/mol) 293.32
Synonym 4,5-diphenyl-2-Oxazolepropanoic acid, 2-Oxazolepropionic acid, 4,5-diphenyl- (8CI), 3-(4,5-Diphenyloxazol-2-yl)propionic acid, 4,5-Diphenyl-2-oxazolepropionic acid, Actirin, Alvo, Daypro, Durapro, Duraprost, NSC 310839, Oxapro, Oxaprozin, Wy 21743, β-(4,5-Diphenyloxazol-2-yl)propionic acid
IUPAC Name 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid
SMILES OC(=O)CCc1oc(c2ccccc2)c(n1)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H15NO3/c20-16(21)12-11-15-19-17(13-7-3-1-4-8-13)18(22-15)14-9-5-2-6-10-14/h1-10H,11-12H2,(H,20,21)
Formula Weight 293.1052
Chemical Name or Material Oxaprozin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.