missing translation for 'onlineSavingsMsg'
Learn More

Oxcarbazepine, MedChemExpress

Product Code. 16478348
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478348 10 mg 10mg
16404247 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478348 Supplier Medchem Express Supplier No. HYB0114
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxcarbazepine is a sodium channel blocker. Oxcarbazepine significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines. Anti-cancer and anticonvulsant effects.

Oxcarbazepine is a sodium channel blocker. Oxcarbazepine significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines. Anti-cancer and anticonvulsant effects.

Specifications

Chemical Name or Material Oxcarbazepine
Physical Form Solid
CAS 28721-07-5
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H12N2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym GP 47680
Solubility Information DMSO : 50 mg/mL (198.20 mM; Need ultrasonic)
SMILES O=C(N1C2=CC=CC=C2CC(C3=CC=CC=C31)=O)N
Molecular Weight (g/mol) 252.27
Formula Weight 252.27
Percent Purity 98.14%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.