missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXCT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98535-50ul
This item is not returnable.
View return policy
Description
OXCT1 Polyclonal antibody specifically detects OXCT1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| OXCT1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| 3-oxoacid CoA transferase 1,3-oxoacid CoA transferase, EC 2.8.3, EC 2.8.3.5, SCOTmitochondrial | |
| Produced in rabbits immunized with E. coli-derived Human OXCT1 fragment. | |
| 50 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 5019 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction