missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OXCT1 Recombinant Mouse Monoclonal Antibody (KBL001_10D10)
GREENER_CHOICE

Product Code. 30827042
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30827042 100 μL -
30830336 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827042 Supplier Invitrogen™ Supplier No. MA564490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: TKFYTDPVEA VKDIPDGATV LVGGFGLCGI PENLIDALLK TGVKGLTAVS NNAGVDNFGL GLLLRSKQIK RMVSSYVGEN AEFERQYLSG ELEVELTPQG TLAERIRAGG AGVPAFYTPT GYGTLVQEGG SPIKYNKDGS VAIASKPREV REFNGQHFIL EEAITGDFAL VKAWKADRAG NVIFRKSARN FNLPMCKAAE TTVVEVEEIV DIGAFAPEDI HIPQIYVHRL IKGEKYEKRI ERLSIRKEGD GEAKSAKPGD DVRERIIKRA ALEFEDGMYA NLGIGIPLLA SNFISPNITV HLQSENGVLG LGPYPRQHEA DADLINAGKE TVTILPGASF FSSDESFAMI RGGHVDLTML GAMQVSKYGD LANWMIPGKM VKGMGGAMDL VSSAKTKVVV TMEHSAKGNA HKIMEKCTLP LTGKQCVNRI ITEKAVFDVD KKKGLTLIEL WEGLTVDDVQ KSTGCDFAVS PKLMPMQQIA N Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%, Pig - 92%, Cynomolgus monkey - 99%.

This gene encodes a member of the 3-oxoacid CoA-transferase gene family. The encoded protein is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate. Mutations in this gene are associated with succinyl CoA:3-oxoacid CoA transferase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXCT1
Applications Immunoprecipitation
Classification Recombinant Monoclonal
Clone KBL001_10D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene OXCT1
Gene Accession No. P55809
Gene Alias 2610008O03Rik; 3-oxoacid CoA transferase 1; 3-oxoacid CoA-transferase 1; 3-oxoacid-CoA transferase 1; EMBL:AAI66478.1}; OXCT; Oxct1; oxct1 {ECO:0000312; Oxct2a; SCOT; SCOT-s; somatic-type succinyl CoA:3-oxoacid CoA-transferase; somatic-type succinyl-CoA:3-oxoacid CoA-transferase; somatic-type succinyl-CoA:3-oxoacid-CoA-transferase; succinyl CoA:3-oxoacid CoA transferase; succinyl-CoA:3-ketoacid CoA transferase; succinyl-CoA:3-ketoacid coenzyme A transferase 1, mitochondrial; succinyl-CoA:3-ketoacid-CoA transferase; succinyl-CoA:3-ketoacid-coenzyme A transferase 1, mitochondrial; Succinyl-CoA:3-ketoacid-coenzyme A transferase 1, mitochondrial-like protein; Unknown (protein for MGC:137519)
Gene Symbols OXCT1
Host Species Mouse
Immunogen Recombinant human OXCT1 (aa 40-520).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5019
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.