missing translation for 'onlineSavingsMsg'
Learn More

Oxibendazole, MedChemExpress

Product Code. 16448226
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448226 100 mg 100mg
16444327 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448226 Supplier Medchem Express Supplier No. HYB0299
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxibendazole is an effective benzimidazole anthelmintic and is against nema-tode infections. Oxibendazole can induces apoptosis and has anti-cancer and anti-inflammation activities.

Oxibendazole is an effective benzimidazole anthelmintic and is against nema-tode infections. Oxibendazole can induces apoptosis and has anti-cancer and anti-inflammation activities.

Specifications

Chemical Name or Material Oxibendazole
Physical Form Powder
CAS 20559-55-1
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H15N3O3
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 2.5 mg/mL (10.03 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.01 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(OC)NC1=NC2=CC=C(OCCC)C=C2N1
Molecular Weight (g/mol) 249.27
Formula Weight 249.27
Percent Purity 98.91%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.