Learn More
Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial
Make colony counting simple for aerobic bacteria, yeast, mold, coliform bacteria and Enterobacteriaceae with the Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial.
36.80 € - 44.35 €
Specifications
Content And Storage | 15°C to 25°C |
---|---|
Format | Dip Slide |
Product Type | Sample Collection |
Quantity | 10/Pk. |
Product Code | Brand | Description | Quantity | Manufacturing Origin | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Manufacturing Origin | Price | Quantity & Availability | |||
11443959
|
Thermo Scientific™
DS0166A |
Dip Slide Plate Count Agar and MacConkey Agar No. 3 | 10/Pk. | United Kingdom |
38.30 €
Pack of 10 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||
11453959
|
Thermo Scientific™
DS0167A |
Dip Slide Plate Count Agar and MacConkey Agar No. 3 (with added germicide inhibitors) | 10/Pk. | United Kingdom |
38.35 €
Pack of 10 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||
11483959
|
Thermo Scientific™
DS0170A |
Dip Slide Plate Count Agar with OGYE Agar | 10/Pk. | United Kingdom |
38.35 €
Pack of 10 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||
11463959
|
Thermo Scientific™
DS0168A |
Dip Slide Plate Count Agar and Violet Red Bile Glucose Agar | 10/Pk. | United Kingdom |
36.80 €
Pack of 10 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||
11423959
|
Thermo Scientific™
DS0147A |
Dip Slide TTC (Red Spot) | 10/Pk. | United Kingdom |
44.35 €
Pack of 10 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||
11433959
|
Thermo Scientific™
DS0155A |
Dip Slide TTC (Red Spot) and Malt Extract Agar | 10/Pk. | United Kingdom |
38.95 €
Pack of 10 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||
11473959
|
Thermo Scientific™
DS0169A |
Dip Slide Plate Count Agar and Violet Red Bile Glucose Agar (with added germicide inhibitors) | 10/Pk. | United Kingdom |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Make colony counting simple for aerobic bacteria, yeast, mold, coliform bacteria and Enterobacteriaceae with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial. Simple-to-use for the operator and where laboratory facilities are limited, the Dip Slide is coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective.
Originally designed to determine bacterial counts on urine specimens, dip slides have been modified over the years by making the device double-sided, with one side a medium for Gram positive species and the other side a medium for Gram negatives. With these dip slides, the process for colony counting is time- and cost-effective, allowing exposure of the specimens to two different media simultaneously.- Slide is made of easily disposable plastic with raised edges to ensure an even thickness of culture medium
- Molded grid allows for easier colony counting
- Slide is shaped to ensure drainage of fluid from the slide
- Cap forms a convenient handle by which it may be held without risk of touching the culture medium
- Bacteria, fungal and yeast charts are included, make examining growth simple for the operator and where laboratory facilities are limited
Specifications
15°C to 25°C | |
Dip Slide | |
Sample Collection | |
10/Pk. |