Learn More
Thermo Scientific™ Oxoid™ Prepared MacConkey Broth Bottled Media
Product Code. 12593936
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
12593936 100 mL Pack of 10
1 options
Product Code. 12593936 Supplier Thermo Scientific™ Supplier No. BO1124M
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Oxoid™ Prepared MacConkey Broth Bottled Media, 100 mL, containing neutral red for the detection of coliform organisms in water and milk examination.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microbiology Media
Quantity 100 mL