missing translation for 'onlineSavingsMsg'
Learn More

Oxybenzone, MedChemExpress

Product Code. 16480718
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480718 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16480718 Supplier Medchem Express Supplier No. HYA0067
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxybenzone (Benzophenone 3) is a commonly used UV filter in sun tans and skin protectants. Oxybenzone act as endocrine disrupting chemicals (EDCs) and can pass through the placental and blood-brain barriers. Benzophenone-3 impairs autophagy, alters epigenetic status, and disrupts retinoid X receptor signaling in apoptotic neuronal cells.

Oxybenzone (Benzophenone 3) is a commonly used UV filter in sun tans and skin protectants. Oxybenzone act as endocrine disrupting chemicals (EDCs) and can pass through the placental and blood-brain barriers. Benzophenone-3 impairs autophagy, alters epigenetic status, and disrupts retinoid X receptor signaling in apoptotic neuronal cells.

Specifications

Chemical Name or Material Oxybenzone
CAS 131-57-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C14H12O3
Quantity 5 g
Synonym Benzophenone 3
Solubility Information DMSO : 100 mg/mL (438.14 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.38 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=CC=C(OC)C=C1O)C2=CC=CC=C2
Molecular Weight (g/mol) 228.24
Formula Weight 228.24
Percent Purity 99.84%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.