missing translation for 'onlineSavingsMsg'
Learn More

Oxypurinol, MedChemExpress

Product Code. 16493787
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16493787 Supplier Medchem Express Supplier No. HY19657
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Oxipurinol (Oxipurinol), the major active metabolite of Allopurinol, is an inhibitor of xanthine oxidase. Oxipurinol can be used to regulate blood urate levels and treat gout.

Oxipurinol (Oxipurinol), the major active metabolite of Allopurinol, is an inhibitor of xanthine oxidase. Oxipurinol can be used to regulate blood urate levels and treat gout.

Specifications

Chemical Name or Material Oxypurinol
Physical Form Powder
CAS 2465-59-0
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C5H4N4O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym Oxipurinol
Solubility Information DMSO : 12.5 mg/mL (82.18 mM; Need ultrasonic)
SMILES O=C1C2=C(NN=C2)NC(N1)=O
Molecular Weight (g/mol) 152.11
Formula Weight 152.11
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.