missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxytetracycline, 'can Be Used as Secondarystandard'

Product Code. 10742861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
Pakningsstørrelse:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging Pakningsstørrelse
10742861 500 mg Glass Bottle 500mg
11976901 2.5 g Glass Bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10742861 Leverandør Thermo Scientific Chemicals Leverandørnr. 257705000
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 79-57-2
Molecular Formula C22H26N2Na2O11
Molecular Weight (g/mol) 540.43
MDL Number MFCD00003700
InChI Key OLRBUPVCLOPDOT-GETZKYKJSA-M
PubChem CID 54675779
IUPAC Name (4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide
SMILES O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O

Specifications

Type Oxytetracycline
Physical Form Crystalline Powder
Melting Point 183.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation − 223.00
Color Beige to Yellow
Quantity 500 mg
Solubility Information Solubility in water: 0.2g/L. Other solubilities: soluble in acids and alkalies, 25 mg/mL methanol, slightly soluble in acetone and chloroform, practically insoluble in ether
Formula Weight 460.43
Percent Purity ≥80 wt% (as is, calculated) (HPLC)
Chemical Name or Material Oxytetracycline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Oxytetracycline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.