missing translation for 'onlineSavingsMsg'
Learn More

Oxytetracycline Hydrochloride, MP Biomedicals™

Product Code. 17600877 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17600877 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17600877 Supplier MP Biomedicals Supplier No. 0215014510
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synonym: Alamycin; 5-Hydroxytetracycline Hydrochloride, Bisolvomycin, Hydrocyclin, NSC 9169, Teramycin Hydrochloride, Tetramine, Tetran Hydrochloride

Oxytetracycline is a tetracycline analog isolated from the actinomycete streptomyces rimosus. It is an antibiotic indicated for treatment of infections caused by Gram positive and Gram negative microorganisms, such as Mycoplasma pneumoniae, Pasteurella pestis, Escherichia coli, Haemophilus influenzae, and Diplococcus pneumoniae. Oxytetracycline hydrochloride is used in studies on the oxytetracycline-resistance gene (otrA), to study phagosome-lysosome (P-L) fusion in P388D1 cells, and antibiotic susceptibilities of Mycopasma bovis isolates. Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use.

Specifications

CAS 2058-46-0
Color Yellow
Quantity 10 g
compliance-icons compliance-icons
Product Identifier
  • Oxytetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Oxytetracycline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.